Learn More
2-Chloro-4-fluorobenzonitrile, 98%
Product Code. 11472847
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11472847 5 g 5g
11482847 25 g 25g
2 options
Product Code. 11472847
Brand: Thermo Scientific Alfa Aesar A15478.06
Request Bulk or Custom Format
CAS: 60702-69-4 | C7H3ClFN | 155.56 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60702-69-4
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.56
MDL Number MFCD00042523
InChI Key PGKPNNMOFHNZJX-UHFFFAOYSA-N
Synonym 4-fluoro-2-chlorobenzonitrile, benzonitrile, 2-chloro-4-fluoro, 2-chloro-4-fluoro benzonitrile, 2-chloro-4-fluoro-benzonitrile, 2-chloro-4-fluorobenzenecarbonitrile, ncr bg df, pubchem3422, acmc-209mll, 2-chloro-4fluorobenzonitrile, 2-chloro4-fluorobenzonitrile
PubChem CID 109000
IUPAC Name 2-chloro-4-fluorobenzonitrile
SMILES FC1=CC=C(C#N)C(Cl)=C1
Specifications
Melting Point 62°C to 66°C
Quantity 5 g
UN Number UN3439
Beilstein 6323034
Formula Weight 155.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluorobenzonitrile