accessibility menu, dialog, popup

UserName

CAS RN 60702-69-4

FNCl

CAS RN 60702-69-4

IUPAC Name: 2-chloro-4-fluorobenzonitrile
Synonyms: 2-chloro-4-fluorobenzonitrile
Molecular Weight (g/mol): 155.56
Molecular Formula: C7H3ClFN
InChi Key: PGKPNNMOFHNZJX-UHFFFAOYSA-N
SMILES: FC1=CC=C(C#N)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results