Learn More
2-Chloro-3-nitropyridine, TRC
Product Code. 30361526
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361526 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30361526 Supplier Toronto Research Chemicals Supplier No. TRCC59705810G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-3-nitropyridine
CAS 5470-18-8
Molecular Formula C5H3ClN2O2
Quantity 10 g
InChI Formula InChI=1S/C5H3ClN2O2/c6-5-4(8(9)10)2-1-3-7-5/h1-3H
SMILES [O-][N+](=O)c1cccnc1Cl
IUPAC Name 2-chloro-3-nitropyridine
Molecular Weight (g/mol) 158.54
Formula Weight 157.9883