missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-cyanopyridine, TRC

Product Code. 30368429
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368429 50 g 50g
30360599 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30368429 Supplier Toronto Research Chemicals Supplier No. TRCC36511050G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-3-cyanopyridine
CAS 6602-54-6
Molecular Formula C6 H3 Cl N2
Quantity 50 g
Synonym 2-Chloro-3-pyridinecarbonitrile, Nicotinonitrile, 2-chloro- (6CI,7CI,8CI), 2-Chloro-3-cyanopyridine, 2-Chloro-3-pyridinecarbonitrile, 2-Chloronicotinonitrile, 3-Cyano-2-chloropyridine, NSC 73287
InChI Formula InChI=1S/C6H3ClN2/c7-6-5(4-8)2-1-3-9-6/h1-3H
SMILES Clc1ncccc1C#N
IUPAC Name 2-chloropyridine-3-carbonitrile
Molecular Weight (g/mol) 138.55
Formula Weight 137.9985
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.