Learn More
2-Chloro-3-cyanopyridine, 98%
Product Code. 11462567
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462567 1 g 1g
11472567 5 g 5g
11482567 25 g 25g
11349639 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11462567 Supplier Thermo Scientific Alfa Aesar Supplier No. A14818.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 6602-54-6 | C6H3ClN2 | 138.554 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6602-54-6
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.554
MDL Number MFCD00014628
InChI Key JAUPUQRPBNDMDT-UHFFFAOYSA-N
Synonym 2-chloro-3-cyanopyridine, 2-chloronicotinonitrile, 2-chloro-3-pyridinecarbonitrile, 3-cyano-2-chloropyridine, 3-pyridinecarbonitrile, 2-chloro, 2-chloro-nicotinonitrile, 2-chloronicotinitrile, 2-chloro-3-cyano-pyridine, 2-chloro-3-cyanopyridine 2-chloronicotinonitrile, 2chloronicotinonitrile
PubChem CID 81079
IUPAC Name 2-chloropyridine-3-carbonitrile
SMILES C1=CC(=C(N=C1)Cl)C#N
Specifications
Melting Point 105°C to 109°C
Boiling Point 112°C (1 mmHg)
Quantity 1 g
UN Number UN3439
Beilstein 115772
Formula Weight 138.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-3-cyanopyridine