missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylpentane, TRC

Product Code. 30360118
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30360118 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30360118 Supplier Toronto Research Chemicals Supplier No. TRCC384268250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4325-48-8
Molecular Formula C6H13Cl
Quantity 250 mg
InChI Formula InChI=1S/C6H13Cl/c1-4-5-6(2,3)7/h4-5H2,1-3H3
SMILES CCCC(C)(C)Cl
IUPAC Name 2-chloro-2-methylpentane
Molecular Weight (g/mol) 120.62
Formula Weight 120.0706
Chemical Name or Material 2-Chloro-2-methylpentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.