Learn More
2-Chloro-1,3-propanediol, TRC
Product Code. 18070730
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18070730 50 mg 50mg
18092720 100 mg 100mg
18030410 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18070730 Supplier Toronto Research Chemicals Supplier No. TRCC37969050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 497-04-1
Molecular Formula C3 H7 Cl O2
Molecular Weight (g/mol) 110.54
Synonym 2-Chloro-1,3-propanediol, 2-Chloro-1,3-dihydroxypropane, 2-Chloro-1,3-propanediol, 2-Monochloropropane-1,3-diol, Glycerol β-chlorohydrin
IUPAC Name 2-chloropropane-1,3-diol
SMILES OCC(Cl)CO
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C3H7ClO2/c4-3(1-5)2-6/h3,5-6H,1-2H2
Formula Weight 110.0135
Percent Purity >95%
Chemical Name or Material 2-Chloro-1,3-propanediol
Grade GC