Learn More
2-Chloro-1,3-propanediol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30309189
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309189 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30309189 Supplier Dr. Ehrenstorfer Supplier No. DREA11502620AL100
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-1,3-propanediol
CAS 497-04-1
Analyte or Component Names 2-Chloro-1,3-propanediol
Shipping Condition Room Temperature
Molecular Formula C3 H7 Cl O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2-Chloro-1,3-propanediol, 2-Chloro-1,3-dihydroxypropane, 2-Chloro-1,3-propanediol, 2-Monochloropropane-1,3-diol, Glycerol β-chlorohydrin, Glycerol Beta-Chlorohydrin, 2CPD, 2-MCPD
InChI Formula InChI=1S/C3H7ClO2/c4-3(1-5)2-6/h3,5-6H,1-2H2
SMILES OCC(Cl)CO
IUPAC Name 2-chloropropane-1,3-diol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 110.54
Quantity 1 mL
Formula Weight 110.0135 g/mol