Learn More
2-Butyl-5-nitrobenzofuran, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Butyl-5-nitrobenzofuran |
| CAS | 133238-87-6 |
| Molecular Formula | C12H13NO3 |
| Quantity | 1 g |
| Synonym | 2-n-Butyl-5-nitrobenzofuran |
| InChI Formula | InChI=1S/C12H13NO3/c1-2-3-4-11-8-9-7-10(13(14)15)5-6-12(9)16-11/h5-8H,2-4H2,1H3 |
| SMILES | CCCCc1oc2ccc(cc2c1)[N+](=O)[O-] |
| IUPAC Name | 2-butyl-5-nitro-1-benzofuran |
| Molecular Weight (g/mol) | 219.24 |
| Formula Weight | 219.0895 |