missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 15487725
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487725 250 mg 250mg
11333029 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487725 Supplier Thermo Scientific Alfa Aesar Supplier No. H27813.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in medicines.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458532-82-6
Molecular Formula C11H15BBrNO2
Molecular Weight (g/mol) 283.96
MDL Number MFCD06798252
InChI Key NQVRUHXWNFMWQB-UHFFFAOYSA-N
Synonym 2-bromo-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-4-boronic acid pinacol ester, 2-bromo-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-bromo-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, ksc495i6f, 2-bromopyridine-4-boronic acid, pinacol ester, 2-bromopyridin-4-ylboronic acid pinacol ester, 2-bromopyridin-4-yl boronic acid pinacol ester, 2-2-bromo 4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-bromo-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 12060163
IUPAC Name 2-bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=NC=C2)Br

Specifications

Melting Point 80°C to 83°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 283.96
Percent Purity 97%
Chemical Name or Material 2-Bromopyridine-4-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.