Learn More
2-Bromopyridine-4-boronic acid, 95%
Product Code. 15473727
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15473727 250 mg 250mg
11416520 1 g 1g
2 options
Product Code. 15473727
Brand: Thermo Scientific Alfa Aesar H27599.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 458532-94-0 | C5H5BBrNO2 | 201.81 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 458532-94-0
Molecular Formula C5H5BBrNO2
Molecular Weight (g/mol) 201.81
MDL Number MFCD06798244
InChI Key DFIKFQMGZNTFLI-UHFFFAOYSA-N
Synonym 2-bromopyridine-4-boronic acid, 2-bromopyridin-4-yl boronic acid, boronic acid, 2-bromo-4-pyridinyl-9ci, 2-bromo-4-pyridyl boronic acid, 2-bromo-4-pyridineboronic acid, boronicacid, 2-bromo-4-pyridinyl-9ci, acmc-209k4o, ksc234s1f, 2-bromo-4-pyridinyl boronic acid, 2-bromanylpyridin-4-yl boronic acid
PubChem CID 12060162
IUPAC Name (2-bromopyridin-4-yl)boronic acid
SMILES OB(O)C1=CC(Br)=NC=C1
Specifications
Quantity 250 mg
Formula Weight 201.82
Percent Purity 95%
Chemical Name or Material 2-Bromopyridine-4-boronic acid