missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenoxyacetonitrile, 99%, Thermo Scientific™

Product Code. 11483957
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483957 2.5 g 2.5g
11493957 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483957 Supplier Thermo Scientific Alfa Aesar Supplier No. A19354.88
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90004-90-3
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
MDL Number MFCD00798567
InChI Key VNMBGABMQPZLCO-UHFFFAOYSA-N
Synonym 2-2-bromophenoxy acetonitrile, 2-bromophenoxyacetonitrile, 2-2-bromophenoxy-acetonitrile, 2-2-bromophenoxy ethanenitrile, acmc-209r4a, 2-o-bromophenoxy acetonitrile
PubChem CID 2734208
IUPAC Name 2-(2-bromophenoxy)acetonitrile
SMILES C1=CC=C(C(=C1)OCC#N)Br

Specifications

Melting Point 47°C to 51°C
Quantity 2.5 g
UN Number UN3439
Formula Weight 212.05
Percent Purity 99%
Chemical Name or Material 2-Bromophenoxyacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.