Learn More
2-Bromoethylammonium Bromide, Sigma-Aldrich™
Product Code. 15836630
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15836630 100 g 100g
15800801 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15836630 Supplier Sigma Aldrich Supplier No. 8201760100
 Restricted Item
Estimated Shipment: 18-12-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2576-47-8
Grade Research
Molecular Weight (g/mol) 204.89
Specifications
Melting Point 169°C to 176°C
MDL Number MFCD00012886
Quantity 100 g
Synonym 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide, 2-Bromoethylammonium bromide