Learn More
(2-Bromoethyl)trimethylammonium Bromide, TRC
Product Code. 30342192
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342192 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30342192 Supplier Toronto Research Chemicals Supplier No. TRCB68434550G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material (2-Bromoethyl)trimethylammonium bromide
CAS 2758-06-7
Molecular Formula C5 H13 Br N . Br
Quantity 50 g
Synonym Bromocholine Bromide
InChI Formula InChI=1S/C5H13BrN.BrH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1H/q+1;/p-1
SMILES [Br-].C[N+](C)(C)CCBr
IUPAC Name 2-bromoethyl(trimethyl)azanium;bromide
Molecular Weight (g/mol) 246.97
Formula Weight 244.9415