Learn More
2-Bromobornane, technical
Product Code. 15567529 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15567529 25 g 25g
15557529 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15567529 Supplier Thermo Scientific Acros Supplier No. 460740250
Request Bulk or Custom Format
CAS: 4443-48-5 | C10H17Br | 217.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4443-48-5
Molecular Formula C10H17Br
Molecular Weight (g/mol) 217.15
InChI Key OTOQMOVZIUGCQE-UHFFFAOYSA-N
Synonym bornyl bromide, 2-bromobornane, bornyl bromide, pract., 2-bromo-1,7,7-trimethylbicyclo 2.2.1 heptane, bornylbromid, bornylbromide, bornane, 2-bromo, 3-bromo-4,7,7-trimethylbicyclo 2.2.1 heptane, 2-bromo-1,7,7-trimethylbicyclo 2.2.1 heptane #
PubChem CID 138249
IUPAC Name 3-bromo-4,7,7-trimethylbicyclo[2.2.1]heptane
SMILES CC1(C2CCC1(C(C2)Br)C)C
Specifications
Boiling Point 94.0°C to 96.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 217.15
Percent Purity ≥78.0% (GC)
Grade Technical
Chemical Name or Material 2-Bromobornane