accessibility menu, dialog, popup

UserName

CAS RN 4443-48-5

H3CH3CCH3Br

CAS RN 4443-48-5

IUPAC Name: 2-bromo-1,7,7-trimethylbicyclo[2.2.1]heptane
Synonyms: 2-bromo-1,7,7-trimethylbicyclo[2.2.1]heptane
Molecular Weight (g/mol): 217.15
Molecular Formula: C10H17Br
InChi Key: OTOQMOVZIUGCQE-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(Br)C2

 No Refinements