Learn More
2-Bromo-5-chlorobenzotrifluoride, 98+%, Thermo Scientific™
Product Code. 11394586
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394586 5 g 5g
11304596 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11394586 Supplier Thermo Scientific Alfa Aesar Supplier No. L20135.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 344-65-0
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00010308
InChI Key OSTIALFVJOFNPP-UHFFFAOYSA-N
Synonym 2-bromo-5-chlorobenzotrifluoride, 1-bromo-4-chloro-2-trifluoromethyl benzene, benzene, 1-bromo-4-chloro-2-trifluoromethyl, 2-bromo-5-chorobenzotrifluoride, 2-bromo-5-chloro-alpha,alpha,alpha-trifluorotoluene, 1-bromo-4-chloro-2-trifluoromethyl-benzene, 4-chloro-2-trifluoromethyl bromobenzene, pubchem2713, bromo-5-chlorobenzotrifluoride
PubChem CID 136172
IUPAC Name 1-bromo-4-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)Br
Spezifikation
Melting Point 18°C to 20°C
Density 1.752
Boiling Point 198°C
Refractive Index 1.505
Quantity 5 g
Beilstein 2448030
Formula Weight 259.44
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-chlorobenzotrifluoride