missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-4'-methylacetanilide, 98%, Thermo Scientific™

Product Code. 11495836
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495836 25 g 25g
11485836 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495836 Supplier Thermo Scientific Alfa Aesar Supplier No. B22213.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-83-5
Molecular Formula C9H10BrNO
Molecular Weight (g/mol) 228.089
MDL Number MFCD00017781
InChI Key UUDGTWKIIUEVJD-UHFFFAOYSA-N
Synonym n-2-bromo-4-methylphenyl acetamide, 2'-bromo-4'-methylacetanilide, 2-bromo-4-methylacetanilide, acetamide, n-2-bromo-4-methylphenyl, n-acetyl-2-bromo-p-toluidine, acmc-1bdfo, 2'-bromo-p-acetotoluidide, cambridge id 5104354, 4-acetamino-3-bromo-toluene, 2-bromo-4-methyl acetanilide
PubChem CID 69200
IUPAC Name N-(2-bromo-4-methylphenyl)acetamide
SMILES CC1=CC(=C(C=C1)NC(=O)C)Br

Specifications

Melting Point 117°C to 119°C
Quantity 25 g
Beilstein 2691019
Formula Weight 228.09
Percent Purity 98%
Chemical Name or Material 2'-Bromo-4'-methylacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.