Learn More
2-Bromo-3'-nitroacetophenone, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 2227-64-7 |
| Molecular Formula | C8H6BrNO3 |
| MDL Number | MFCD00024512 |
| Quantity | 25 g |
| SMILES | O=[N+](C1=CC(C(CBr)=O)=CC=C1)[O-] |
| Molecular Weight (g/mol) | 244.04 |
| Percent Purity | 96% |
| Chemical Name or Material | 2-Bromo-3'-nitroacetophenone |