Learn More
2-Bromo-3-methylthiophene, 97%
Product Code. 11387746
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11387746 5 g 5g
11397746 25 g 25g
2 options
Product Code. 11387746
Brand: Thermo Scientific Alfa Aesar L11393.06
Request Bulk or Custom Format
CAS: 14282-76-9 | C5H5BrS | 177.059 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14282-76-9
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.059
MDL Number MFCD00059741
InChI Key YYJBWYBULYUKMR-UHFFFAOYSA-N
Synonym thiophene, 2-bromo-3-methyl, 2-bromo-3-methyl-thiophene, 1-bromo-2-methyl thiophene, pubchem5191, 2-bromo-3methylthiophene, 2-bromo-3-methythiophene, acmc-1buh4, 2-bromo-3methyl thiophene, 2-bromo-3-methyl thiophene, ksc491i3r
PubChem CID 84314
IUPAC Name 2-bromo-3-methylthiophene
SMILES CC1=C(SC=C1)Br
Specifications
Density 1.58
Boiling Point 173°C to 176°C
Flash Point 68°C (154°F)
Refractive Index 1.571
Quantity 5 g
UN Number UN2810
Beilstein 105719
Formula Weight 177.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-methylthiophene