Learn More
2-Bromo-3-fluorobenzyl bromide, 98%
Product Code. 15440429
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440429 1 g 1g
15450429 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15440429 Supplier Thermo Scientific Alfa Aesar Supplier No. H63632.03
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
CAS: 1184918-22-6 | C7H5Br2F | 267.92 g/mol
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1184918-22-6
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.92
MDL Number MFCD12911576
InChI Key ZJSRIMJDFLFPJI-UHFFFAOYSA-N
Synonym 2-bromo-1-bromomethyl-3-fluorobenzene, 2-bromo-3-fluorobenzyl bromide, 2-bromo-3-bromomethyl-1-fluorobenzene, 2-bromo-3-fluorobenzylbromide
PubChem CID 44181799
IUPAC Name 2-bromo-1-(bromomethyl)-3-fluorobenzene
SMILES FC1=CC=CC(CBr)=C1Br
Specifications
Color White
Quantity 1 g
UN Number UN3261
Solubility Information Slightly soluble in water.
Formula Weight 267.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-3-fluorobenzyl bromide