missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-(trifluoromethyl)acetophenone, 97%

Product Code. 15454707
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454707 250 mg 250mg
15434707 1 g 1g
15444707 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15454707 Supplier Thermo Scientific Alfa Aesar Supplier No. H32032.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 54109-16-9 | C9H6BrF3O | 267.045 g/mol

The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54109-16-9
Molecular Formula C9H6BrF3O
Molecular Weight (g/mol) 267.045
MDL Number MFCD03094304
InChI Key KWZCBMKXNYOQAK-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenacyl bromide, 2-bromo-1-2-trifluoromethyl phenyl-1-ethanone, 2-bromo-2'-trifluoromethyl acetophenone, 2-bromo-1-2-trifluoromethyl-phenyl-ethanone, 2-bromo-1-2-trifluoromethyl phenyl ethan-1-one, 2-bromo-1-2-trifluoromethyl phenyl ethanone, ethanone, 2-bromo-1-2-trifluoromethyl phenyl, 2-trifluoromethyphenacyl bromide, 2-trifluoromethyl phenacylbromide
PubChem CID 2778430
IUPAC Name 2-bromo-1-[2-(trifluoromethyl)phenyl]ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)C(F)(F)F

Specifications

Melting Point 24°C to 32°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 267.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-2'-(trifluoromethyl)acetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.