missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-cyclopropyl-2-(4-fluorophenyl)ethanone, TRC

Product Code. 30240795
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240795 50 mg 50mg
30240999 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240795 Supplier Toronto Research Chemicals Supplier No. TRCB68278550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1359829-52-9
Molecular Formula C11H10BrFO
Molecular Weight (g/mol) 257.1
IUPAC Name 2-bromo-1-cyclopropyl-2-(4-fluorophenyl)ethanone
SMILES Fc1ccc(cc1)C(Br)C(=O)C2CC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H10BrFO/c12-10(11(14)8-1-2-8)7-3-5-9(13)6-4-7/h3-6,8,10H,1-2H2
Formula Weight 255.9899
Chemical Name or Material 2-Bromo-1-cyclopropyl-2-(4-fluorophenyl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.