Learn More
2-Bromo-1-benzofuran, 97%, Thermo Scientific™
Product Code. p-4812748
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. 10592662
Brand: Thermo Scientific Maybridge CC06610DA
Request Bulk or Custom Format
Chemical Identifiers
CAS 54008-77-4
Molecular Formula C8H5BrO
Molecular Weight (g/mol) 197.031
InChI Key RNEOFIVNTNLSEH-UHFFFAOYSA-N
Synonym 2-bromobenzofuran, 2-bromobenzo b furan, bromobenzofuran, 2-bromo-1-benzofurane, benzofuran, 2-bromo, pubchem13460, 2-bromanyl-1-benzofuran, 2-bromo-1-benzo b furan
PubChem CID 2776264
IUPAC Name 2-bromo-1-benzofuran
SMILES C1=CC=C2C(=C1)C=C(O2)Br
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 197.03
Chemical Name or Material 2-Bromo-1-benzofuran