accessibility menu, dialog, popup

UserName

CAS RN 54008-77-4

OBr

CAS RN 54008-77-4

IUPAC Name: 2-bromo-1-benzofuran
Synonyms: 2-bromo-1-benzofuran
Molecular Weight (g/mol): 197.03
Molecular Formula: C8H5BrO
InChi Key: RNEOFIVNTNLSEH-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results