Learn More
2-Bromo-1-(4-chloro-3-nitrophenyl)ethan-1-one, Technical Grade, Thermo Scientific™
Product Code. 10427622
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10427622 1 g Amber Glass Bottle 1g
10334192 10 g Amber Glass Bottle 10g
10327014 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10427622 Supplier Thermo Scientific Maybridge Supplier No. JP00700DA
 Restricted Item
Estimated Shipment: 15-01-2027
Chemical Identifiers
CAS 22019-49-4
Molecular Formula C8H5BrClNO3
Molecular Weight (g/mol) 278.486
InChI Key FXNQTMRPLLNPQY-UHFFFAOYSA-N
PubChem CID 2778330
IUPAC Name 2-bromo-1-(4-chloro-3-nitrophenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CBr)[N+](=O)[O-])Cl
Specifications
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-bromo-1-4-chloro-3-nitrophenyl ethanone, 2-bromo-1-4-chloro-3-nitrophenyl ethan-1-one, 4-chloro-3-nitrophenacylbromide, 4-chloro-3-nitrophenacyl bromide, ethanone,2-bromo-1-4-chloro-3-nitrophenyl, 4-chloro-3-nitro-phenacyl bromide, 4-chloro-3-nitrophen-acyl bromide, 2-bromo-4'-chloro-3'-nitroacetophenone, 2-bromo-1-4-chloro-3-nitro-phenyl ethanone, ethanone, 2-bromo-1-4-chloro-3-nitrophenyl
Formula Weight 278.49
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-chloro-3-nitrophenyl)ethan-1-one