Learn More
2-Bromo-1-(1,3-thiazol-2-yl)ethanone, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-1-(1,3-thiazol-2-yl)ethanone |
| CAS | 3292-77-1 |
| Molecular Formula | C5H4BrNOS |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C5H4BrNOS/c6-3-4(8)5-7-1-2-9-5/h1-2H,3H2 |
| SMILES | BrCC(=O)c1nccs1 |
| IUPAC Name | 2-bromo-1-(1,3-thiazol-2-yl)ethanone |
| Molecular Weight (g/mol) | 206.06 |
| Formula Weight | 204.9197 |