missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylacrylic Acid, TRC

Product Code. 30136779
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136779 1 g 1g
30134703 10 g 10g
30006610 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136779 Supplier Toronto Research Chemicals Supplier No. TRCB2243801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5669-19-2
Molecular Formula C10 H10 O2
Molecular Weight (g/mol) 162.19
Synonym Hydrocinnamic acid, α-methylene- (6CI,8CI), 2-Benzylacrylic acid, 2-Methylene-3-phenylpropanoic acid, NSC 192640, α-Benzylacrylic acid, α-Methylenebenzenepropanoic acid, 2-Benzylprop-2-enoic acid
IUPAC Name 2-benzylprop-2-enoic acid
SMILES OC(=O)C(=C)Cc1ccccc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H10O2/c1-8(10(11)12)7-9-5-3-2-4-6-9/h2-6H,1,7H2,(H,11,12)
Formula Weight 162.0681
Chemical Name or Material 2-Benzylacrylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.