missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiophene-3-carbonitrile, 97%, Thermo Scientific™

Product Code. p-2697743
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10191162 1 g Amber Glass Bottle 1g
10101172 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10191162 Supplier Thermo Scientific Maybridge Supplier No. GK01096DA
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4651-82-5
Molecular Formula C5H4N2S
Molecular Weight (g/mol) 124.16
MDL Number MFCD00706298
InChI Key XVGHZFWFGXDIOU-UHFFFAOYSA-N
Synonym 2-amino-3-cyanothiophene, 2-amino-3-thiophenecarbonitrile, 2-amino-thiophene-3-carbonitrile, 3-thiophenecarbonitrile,2-amino, 3-thiophenecarbonitrile, 2-amino, 2-aminothiophen-3-carbonitril, pubchem5222, aminothiophenecarbonitrile, 2-amino-3-thienyl cyanide, 2-azanylthiophene-3-carbonitrile
PubChem CID 736607
IUPAC Name 2-aminothiophene-3-carbonitrile
SMILES NC1=C(C=CS1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 124.17
Percent Purity 97%
Chemical Name or Material 2-Aminothiophene-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.