Learn More
2-Amino-n-(tert-butyl)benzenesulfonamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-n-(tert-butyl)benzenesulfonamide |
| CAS | 954268-81-6 |
| Molecular Formula | C10H16N2O2S |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H16N2O2S/c1-10(2,3)12-15(13,14)9-7-5-4-6-8(9)11/h4-7,12H,11H2,1-3H3 |
| SMILES | CC(C)(C)NS(=O)(=O)C1=CC=CC=C1N |
| IUPAC Name | 2-amino-N-tert-butylbenzenesulfonamide |
| Molecular Weight (g/mol) | 228.31 |
| Formula Weight | 228.0932 |