Learn More
2-Amino-N-(3,6-dichloro-2-methylphenyl)thiazole-5-carboxamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-N-(3,6-dichloro-2-methylphenyl)thiazole-5-carboxamide |
| CAS | 2468737-79-1 |
| Molecular Formula | C11H9Cl2N3OS |
| Quantity | 100 mg |
| Synonym | 2-Amino-N-(3,6-dichloro-2-methylphenyl)thiazole-5-carboxamide, Dasatinib Impurity |
| InChI Formula | InChI=1S/C11H9Cl2N3OS/c1-5-6(12)2-3-7(13)9(5)16-10(17)8-4-15-11(14)18-8/h2-4H,1H3,(H2,14,15)(H,16,17) |
| SMILES | NC1=NC=C(C(NC2=C(Cl)C=CC(Cl)=C2C)=O)S1 |
| IUPAC Name | 2-amino-N-(3,6-dichloro-2-methylphenyl)thiazole-5-carboxamide |
| Molecular Weight (g/mol) | 302.18 |
| Formula Weight | 300.98 |