missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-(trifluoromethoxy)phenyl Thiocyanate, TRC

Product Code. 30340177
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340177 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340177 Supplier Toronto Research Chemicals Supplier No. TRCA630770500MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-5-(trifluoromethoxy)phenyl Thiocyanate
CAS 1391054-04-8
Molecular Formula C8 H5 F3 N2 O S
Quantity 500 mg
Synonym Thiocyanic acid, 2-amino-5-(trifluoromethoxy)phenyl ester, [2-Amino-5-(trifluoromethoxy)phenyl] Thiocyanate
InChI Formula InChI=1S/C8H5F3N2OS/c9-8(10,11)14-5-1-2-6(13)7(3-5)15-4-12/h1-3H,13H2
SMILES Nc1ccc(OC(F)(F)F)cc1SC#N
IUPAC Name [2-amino-5-(trifluoromethoxy)phenyl] thiocyanate
Molecular Weight (g/mol) 234.2
Formula Weight 234.0075
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.