missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzeneboronic acid pinacol ester, 96%

Product Code. p-7041558
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497586 250 mg 250mg
15487586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497586 Supplier Thermo Scientific Alfa Aesar Supplier No. H62575.MD
Add to basket
This item is not returnable. View return policy

CAS: 863578-24-9 | C12H17BFNO2 | 237.081 g/mol

2-Amino-5-fluorobenzeneboronic acid pinacol ester is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-fluorobenzeneboronic acid pinacol ester is used as a pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863578-24-9
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.081
MDL Number MFCD11617916
InChI Key RLUKWTHMONYTKG-UHFFFAOYSA-N
Synonym 4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-amino-5-fluorophenyl boronic acid pinacol ester, benzenamine,4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-amino-5-fluorobenzeneboronic acid pinacol ester, 4-fluoro-2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-fluoranyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine, 4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 4-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 23152508
IUPAC Name 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)F)N

Specifications

Melting Point 49°C to 50°C
Quantity 250 mg
Formula Weight 237.08
Percent Purity 96%
Chemical Name or Material 2-Amino-5-fluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.