Learn More
2-Amino-5,5-dimethyl-5,6-dihydro-4h-benzothiazol-7-one, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-5,5-dimethyl-5,6-dihydro-4h-benzothiazol-7-one |
| CAS | 1629-95-4 |
| Molecular Formula | C9H12N2OS |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C9H12N2OS/c1-9(2)3-5-7(6(12)4-9)13-8(10)11-5/h3-4H2,1-2H3,(H2,10,11) |
| SMILES | CC1(C)CC(=O)c2sc(N)nc2C1 |
| IUPAC Name | 2-amino-5,5-dimethyl-4,6-dihydro-1,3-benzothiazol-7-one |
| Molecular Weight (g/mol) | 196.27 |
| Formula Weight | 196.067 |