missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloropyrimidine, 98%

Product Code. 15481118
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481118 250 mg 250mg
15461118 1 g 1g
15471118 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481118 Supplier Thermo Scientific Alfa Aesar Supplier No. H55007.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3993-78-0 | C4H4ClN3 | 129.547 g/mol

2-Amino-4-chloropyrimidine is a substrate used in a palladium-catalyzed cyanation with zinc(II) cyanide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-chloropyrimidine is a substrate used in a palladium-catalyzed cyanation with zinc(II) cyanide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3993-78-0
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD00038021
InChI Key DBGFGNCFYUNXLD-UHFFFAOYSA-N
Synonym 2-amino-4-chloropyrimidine, 2-amine-4-chloropyrimidine, 2-pyrimidinamine, 4-chloro, 4-chloro-2-pyrimidinamine, 4-chloro-2-aminopyrimidine, 4-chloropyrimidine-2-amine, 4-chloro-pyrimidin-2-ylamine, 4-chloropyrimidine-2-ylamine, 2-amino-6-chloropyrimidine, 4-chloro-2-pyrimidineamine
PubChem CID 223332
IUPAC Name 4-chloropyrimidin-2-amine
SMILES C1=CN=C(N=C1Cl)N

Specifications

Melting Point 155°C to 160°C (decomposition)
Quantity 250 mg
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 129.55
Percent Purity 98%
Chemical Name or Material 2-Amino-4-chloropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.