Learn More
2-Amino-3-cyanopyridine, 98%
Product Code. 11325395
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325395 5 g 5g
11315395 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11325395 Supplier Thermo Scientific Alfa Aesar Supplier No. L19618.06
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
CAS: 24517-64-4 | C6H6N3 | 120.13 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24517-64-4
Molecular Formula C6H6N3
Molecular Weight (g/mol) 120.13
MDL Number MFCD00128867
InChI Key YYXDQRRDNPRJFL-UHFFFAOYSA-O
Synonym 2-amino-3-cyanopyridine, 2-aminonicotinonitrile, 2-amino-3-pyridinecarbonitrile, 2-amino-pyridine-3-carbonitrile, 2-amino-nicotinonitrile, 3-pyridinecarbonitrile, 2-amino, 3-cyanopyridin-2-amine, 3-cyanopyridine-2-amine, aminonicotinonitrile, zlchem 506
PubChem CID 818259
IUPAC Name 2-amino-3-cyanopyridin-1-ium
SMILES NC1=[NH+]C=CC=C1C#N
Specifications
Melting Point 133°C to 135°C
Quantity 5 g
UN Number UN3439
Beilstein 115612
Formula Weight 119.13
Percent Purity 98%
Chemical Name or Material 2-Amino-3-cyanopyridine