missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,6-dibromopyrazine, 95%, Thermo Scientific™

Product Code. 15415428
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415428 250 mg 250mg
15405428 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415428 Supplier Thermo Scientific Alfa Aesar Supplier No. H59185.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957230-70-5
Molecular Formula C4H3Br2N3
Molecular Weight (g/mol) 252.897
MDL Number MFCD11040377
InChI Key ZEDFFSRZLCREEN-UHFFFAOYSA-N
Synonym 2-amino-3,6-dibromopyrazine, 3,6-dibromopyrazine-2-amine, 3,6-dibromo-2-pyrazinamine, 3-amino-2,5-dibromopyrazine, 3,6-dibromo-pyrazin-2-ylamine, 2,5-dibromo-3-aminopyrazine, 3,6-dibromopyrazin-2-yl amine, 3,6-bis bromanyl pyrazin-2-amine, 2-pyrazinamine, 3,6-dibromo
PubChem CID 51051665
IUPAC Name 3,6-dibromopyrazin-2-amine
SMILES C1=C(N=C(C(=N1)Br)N)Br

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 252.9
Percent Purity 95%
Chemical Name or Material 2-Amino-3,6-dibromopyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.