Learn More
2,8-Dibromodibenzo[b,d]thiophene, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 31574-87-5 |
| Molecular Formula | C12H6Br2S |
| MDL Number | MFCD00092755 |
| Quantity | 500 g |
| SMILES | BrC1=CC=C2C(C3=CC(Br)=CC=C3S2)=C1 |
| Molecular Weight (g/mol) | 342.05 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,8-Dibromodibenzo[b,d]thiophene |