missing translation for 'onlineSavingsMsg'
Learn More

2,6-Naphthalenedisulfonic Acid, Sodium Salt (Technical), MP Biomedicals

Product Code. 11457305 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457305 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11457305 Supplier MP Biomedicals Supplier No. 0520942880

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1655-45-4
Molecular Formula C10H6Na2O6S2
Quantity 100 g
Synonym 2,6-naphthalenedisulfonic acid disodium salt, sodium naphthalene-2,6-disulfonate, disodium 2,6-naphthalenedisulfonate, disodium naphthalene-2,6-disulphonate, 2,6-naphthalenedisulfonic acid, disodium salt, ebert-merz beta-acid disodium salt, disodium naphthalene-2,6-disulfonate, dipotassium 2,6-naphthalenedisulfonate, acmc-209dsq, ksc910s4f
InChI Key WZZLWPIYWZEJOX-UHFFFAOYSA-L
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)[O-])C=C1S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;naphthalene-2,6-disulfonate
Molecular Weight (g/mol) 332.252
PubChem CID 3014027
Formula Weight 332.26
Chemical Name or Material 2,6-Naphthalenedisulfonic Acid, Na Salt, Tech
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.