Learn More
2,6-Dimethylbenzyl chloride, 98%, Thermo Scientific™
Product Code. p-7077679 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15515976 5 g 5g
15525976 25 g 25g
15535976 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15515976 Supplier Thermo Scientific Acros Supplier No. 457470050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 5402-60-8
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.64
InChI Key HPVRFWQMBYLJRL-UHFFFAOYSA-N
Synonym 2,6-dimethylbenzyl chloride, 2-chloromethyl-1,3-dimethylbenzene, 2,6-dimethylbenzylchloride, 2,6-dimethyl benzylchloride, 1-chloromethyl-2,6-dimethylbenzene, 2-chloromethyl-1,3-dimethyl-benzene, 2-chloromethyl-m-xylene, benzene, 2-chloromethyl-1,3-dimethyl, 2, 6-dimethylbenzyl chloride
PubChem CID 221154
IUPAC Name 2-(chloromethyl)-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)CCl
Specifications
Melting Point 33.0°C
Boiling Point 100°C (2.0 kPa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 154.64
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylbenzyl chloride
compliance-icons
Product Identifier
  • 2,6-Dimethylbenzyl chloride, 98%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.