Learn More
2,6-Dimethoxyphenol-3,4,5-d3, CDN
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethoxyphenol-3,4,5-D3 |
| CAS | 2484091-60-1 |
| Molecular Formula | C8 D3 H7 O3 |
| Quantity | 0.25 g |
| Synonym | Phen-3,4,5-d3-ol, 2,6-dimethoxy- (ACI), 2,6-Dimethoxyphen-3,4,5-d3-ol (ACI), 2,6-Dimethoxyphenol-3,4,5-d3, 2,6-Dimethoxyphenol-d3 (phenyl-d3), 2,6-Dimethoxyphenol-3,4,5-D3 |
| InChI Formula | InChI=1 S/C8H10O3/c1-10-6-4-3-5-7(11-2)8(6)9/h3-5,9 H,1-2H3/i3D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(OC)c(O)c(OC)c1[2 H] |
| IUPAC Name | 3,4,5-trideuterio-2,6-dimethoxyphenol |
| Molecular Weight (g/mol) | 157.1817 |
| Formula Weight | 157.0818 g/mol |
| Show More |