missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzoic acid, 98+%

Product Code. p-7021992
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11441948 25 g 25g
11451948 100 g 100g
11461948 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11441948

Brand: Thermo Scientific Alfa Aesar A14558.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 385-00-2 | C7H4F2O2 | 158.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 385-00-2
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00002411
InChI Key ONOTYLMNTZNAQZ-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-difluoro, 2,6-difluorobenzoicacid, 2,6-difluoro benzoic acid, 2,6-difluoro-benzoic acid, pubchem2274, benzoic acid,6-difluoro, acmc-209j0h, 2,6,-difluorobenzoic acid, 3-09-00-01330 beilstein handbook reference, ksc223c7l
PubChem CID 9796
IUPAC Name 2,6-difluorobenzoic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)F

Specifications

Melting Point 156°C to 160°C
Boiling Point 72°C to 77°C (13 mmHg)
Odor Odorless
Quantity 25 g
Beilstein 973774
Formula Weight 158.11
Percent Purity ≥98%
Chemical Name or Material 2,6-Difluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.