accessibility menu, dialog, popup

UserName

CAS RN 385-00-2

HOOFF

CAS RN 385-00-2

IUPAC Name: 2,6-difluorobenzoic acid
Synonyms: benzoic acid, 2,6-difluoro-
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: ONOTYLMNTZNAQZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results