missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-nitrobenzonitrile, 98%, Thermo Scientific™

Product Code. 11347106
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347106 1 g 1g
11357106 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347106 Supplier Thermo Scientific Alfa Aesar Supplier No. L07035.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143879-77-0
Molecular Formula C7H2F2N2O2
Molecular Weight (g/mol) 184.10
MDL Number MFCD00042254
InChI Key JQHDRFKDZHGEBQ-UHFFFAOYSA-N
Synonym benzonitrile, 2,6-difluoro-3-nitro, 2,4-difluoro-3-cyanonitrobenzene, 2,6-difluoro-3-nitrobenzenecarbonitrile, 3-cyano-2,4-difluoronitrobenzene, buttpark 87\01-76, pubchem4771, acmc-209crh, intermediates-zcf02101, ksc495e0t, 2,6-difluoro-3-nitro-benzonitrile
PubChem CID 2737030
IUPAC Name 2,6-difluoro-3-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=C(F)C(C#N)=C1F

Specifications

Melting Point 51°C to 54°C
Quantity 1 g
UN Number UN3439
Beilstein 5431828
Formula Weight 184.1
Percent Purity 98%
Chemical Name or Material 2,6-Difluoro-3-nitrobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.