missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorocinnamic acid, predominantly trans, 98%, Thermo Scientific™

Product Code. p-7044273
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11317515

Brand: Thermo Scientific Alfa Aesar L05614.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5345-89-1
Molecular Formula C9H6Cl2O2
Molecular Weight (g/mol) 217.05
MDL Number MFCD00004374
InChI Key OIPVGRCXMFBNAN-SNAWJCMRSA-N
Synonym 2,6-dichlorocinnamic acid, 3-2,6-dichlorophenyl acrylic acid, e-3-2,6-dichlorophenyl acrylic acid, 2e-3-2,6-dichlorophenyl prop-2-enoic acid, 2e-3-2,6-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,6-dichlorophenyl, 3-2,6-dichlorophenyl-2-propenoic acid, cinnamic acid, 2,6-dichloro-6ci,7ci, 3-2,6-dichlorophenyl propenoic acid, 2,6-dichlorocinnamic acid, predominantly trans
PubChem CID 731762
IUPAC Name (E)-3-(2,6-dichlorophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 193°C to 197°C
Quantity 5 g
Beilstein 1953654
Formula Weight 217.05
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorocinnamic acid, predominantly trans
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.