accessibility menu, dialog, popup

UserName

CAS RN 5345-89-1

HOOClCl(E)

CAS RN 5345-89-1

IUPAC Name: (2E)-3-(2,6-dichlorophenyl)prop-2-enoic acid
Synonyms: (2E)-3-(2,6-dichlorophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 217.05
Molecular Formula: C9H6Cl2O2
InChi Key: OIPVGRCXMFBNAN-SNAWJCMRSA-N
SMILES: OC(=O)\C=C\C1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results