Learn More
2,6-Dibromopyridine, 98%
Product Code. p-7022561
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438356 25 g 25g
11448356 100 g 100g
11458356 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11438356 Supplier Thermo Scientific Alfa Aesar Supplier No. A15397.14
Add to basket
CAS: 626-05-1 | C5H3Br2N | 236.894 g/mol
2,6-Dibromopyridine is used as a tridentate chelating ligand and in the formation of macrocycles containing the terpyridine moiety. It is also used to produce 6-bromo-2-methoxypyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dibromopyridine is used as a tridentate chelating ligand and in the formation of macrocycles containing the terpyridine moiety. It is also used to produce 6-bromo-2-methoxypyridine.
Solubility
Soluble in hot methanol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 626-05-1
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.894
MDL Number MFCD00006223
InChI Key FEYDZHNIIMENOB-UHFFFAOYSA-N
Synonym pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
PubChem CID 12274
IUPAC Name 2,6-dibromopyridine
SMILES C1=CC(=NC(=C1)Br)Br
Specifications
Melting Point 117°C to 120°C
Boiling Point 255°C
Flash Point 213°C (415°F)
Odor Recognizable
Quantity 25 g
UN Number UN2811
Beilstein 108922
Solubility Information Soluble in hot methanol. Insoluble in water.
Formula Weight 236.9
Percent Purity 98%
Chemical Name or Material 2,6-Dibromopyridine