Learn More
2,6,10-Trimethyl-2,6,10-dodecatriene, TRC
Product Code. 30455465
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455465 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455465 Supplier Toronto Research Chemicals Supplier No. TRCT796315100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,6,10-Trimethyl-2,6,10-dodecatriene
CAS 7681-88-1
Molecular Formula C15 H26
Quantity 100 mg
InChI Formula InChI=1S/C15H26/c1-6-14(4)10-8-12-15(5)11-7-9-13(2)3/h6,9,12H,7-8,10-11H2,1-5H3/b14-6+,15-12+
SMILES C\C=C(/C)\CC\C=C(/C)\CCC=C(C)C
IUPAC Name (6E,10E)-2,6,10-trimethyldodeca-2,6,10-triene
Molecular Weight (g/mol) 206.367
Formula Weight 206.203