missing translation for 'onlineSavingsMsg'
Learn More

2-[[(5S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]-1H-isoindole-1,3(2H)-dione, Mikromol™

Product Code. 30050649
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050649 25 mg 25mg
30052133 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050649 Supplier Mikromol Supplier No. MM3300.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[[(5S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]-1H-isoindole-1,3(2H)-dione
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 2-[((5S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl)methyl]-1H-isoindole-1,3(2H)-dione
CAS 168828-89-5
Analyte or Component Names 2-[((5S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl)methyl]-1H-isoindole-1,3(2H)-dione
Shipping Condition Room Temperature
Molecular Formula C22 H20 F N3 O5
Certifications/Compliance ISO 17025
Synonym 1H-Isoindole-1,3(2H)-dione, 2-[[(5S)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]-, 1H-Isoindole-1,3(2H)-dione, 2-[[3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]-, (S)-, (S)-N-[[3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]phthalimide, 2-[[(5S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]-1H-isoindole-1,3(2H)-dione
InChI Formula InChI=1S/C22H20FN3O5/c23-18-11-14(5-6-19(18)24-7-9-30-10-8-24)25-12-15(31-22(25)29)13-26-20(27)16-3-1-2-4-17(16)21(26)28/h1-6,11,15H,7-10,12-13H2/t15-/m1/s1
SMILES Fc1cc(ccc1N2CCOCC2)N3C[C@H](CN4C(=O)c5ccccc5C4=O)OC3=O
IUPAC Name 2-[[(5S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]isoindole-1,3-dione
Molecular Weight (g/mol) 425.41
Quantity 25 mg
Formula Weight 425.1387 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.