Learn More
2,5-Norbornadiene, Sigma-Aldrich™
Specifications
Specifications
| CAS | 121-46-0 |
| Melting Point | -20°C |
| Density | 0.906 g/cm3 at 20°C |
| MDL Number | MFCD00082301 |
| Quantity | 250 mL |
| Synonym | 2,5-Norbornadiene, Bicyclo[2.2.1]hepta-2,5-diene, 2,5-Norbornadiene |
| Molecular Weight (g/mol) | 92.14 |
| Grade | Research |